Läs mer
R&D Systems™ Human Serpin A12 Quantikine ELISA Kit
Kvantitet:
1 sats
Beskrivning
Innehåller
Human Serpin A12 microplate, 96-well; Human Serpin A12 conjugate; Human Serpin A12 standard; Buffers and diluents, Color reagents A and B; Stop solution; Plate sealers
The Human Serpin A12 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Serpin A12 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), saliva (50 ul), urine (50 ul), human milk (50 ul).
Specifikationer
Specifikationer
| Kvantitet | 1 sats |
| Art | Human |
| Konjugera | HRP |
| Analyskänslighet | 14.6 pg/mL |
| Analysintervall | 31.30 to 2000 pg/mL |
| För användning med (applikation) | ELISA |
| Provtyp | Cell Culture Supernates (50uL), Serum (50uL), EDTA Plasma (50uL), Heparin Plasma (50uL), Saliva (50uL), Urine (50uL), Human Milk (50uL) |
| Synonym | antitrypsin), member 12, member 12, OL-64, OL-64Visceral adipose-specific serpin, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serpin A12, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Vaspin, Visceral adipose tissue-derived serine protease inhibitor |
| Innehåll och lagring | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |