Läs mer
R&D Systems™ Human XCL1/Lymphotactin Quantikine ELISA Kit
Kvantitet:
1 sats
Beskrivning
Innehåller
Human Lymphotactin microplate, 96-well; Human Lymphotactin conjugate; Human Lymphotactin standard; Buffers and diluents, Color reagents A and B; Stop solution; Plate sealers
The Human XCL1/Lymphotactin Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human XCL1/Lymphotactin in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul).
Specifikationer
Specifikationer
| Kvantitet | 1 sats |
| Art | Human |
| Konjugera | HRP |
| Analyskänslighet | 0.023 ng/mL |
| Analysintervall | 0.16 to 10 ng/mL |
| För användning med (applikation) | ELISA |
| Provtyp | Cell Culture Supernates (100uL), Serum (100uL), EDTA Plasma (100uL), Heparin Plasma (100uL) |
| Synonym | ATAC, ATACSmall-inducible cytokine C1, C motif chemokine 1, chemokine (C motif) ligand 1, Cytokine SCM-1, LPTNSCM-1-alpha, LTNXC chemokine ligand 1, lymphotactin, lymphotaxin, SCM-1, SCM-1 alpha, SCM-1a, SCM1A, SCYC1SCM1, single cysteine motif 1a, small inducible cytokine subfamily C, member 1 (lymphotactin) |
| Innehåll och lagring | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |