Läs mer
Tocris Bioscience™ KRIBB11
Beskrivning
KRIBB11 is a heat shock factor (HSF) inhibitor (IC50 = 1.2 μM). Increases apoptosis in cancer cells treated with the HSP90 inhibitors Geldanamycin (Cat. No. 1368) and 17-AAG (Cat. No. 1515). Blocks downstream induction of HSP27 and HSP70. Inhibits tumor growth in vivo.
Specifikationer
Specifikationer
| Kemiskt namn eller material | KRIBB11 |
| CAS | 342639-96-7 |
| Synonym | N2-1H-Indazol-5-yl-N6-methyl-3-nitro-2,6-pyridinediamine |
| Kvantitet | 50 mg |
| Löslighet | Soluble to 100mM in DMSO |
| Molekylformel | C13H12N6O2 |
| Formel vikt | 284.27 |
| Fysisk form | Solid |
| Produkttyp | Bioactive Small Molecule |
| Renhet | >99% |
Research Use Only. Not For Human or Veterinary Use