Läs mer
Tocris Bioscience™ ML 193
Potent, selective GPR55 antagonist
Brand: Tocris Bioscience™ 4860/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
ML 193 is a selective GPR55 antagonist (IC50 = 221 nM). Exhibits >27-, >145- and >145-fold selectivity for GPR55 over CB1, GPR35 and CB2, respectively. Inhibits ERK signaling in vitro.Specifikationer
| ML 193 | |
| N-[4-[[(3,4-Dimethyl-5-isoxazolyl)amino]sulfonyl]phenyl]-6,8-dimethyl-2-(2-pyridinyl)-4-quinolinecarboxamide | |
| Soluble to 100mM in DMSO | |
| Bioactive Small Molecule |
| 713121-80-3 | |
| 10 mg | |
| Solid |
Produkttitel
Research Use Only. Not For Human or Veterinary Use