Läs mer
Tocris Bioscience™ NSC 617145
Beskrivning
NSC 617145 is a werner syndrome helicase (WRN) helicase inhibitor (IC50 = 250 nM); acts synergistically with Mitomycin C (Cat. No. 3258) to induce double-strand breaks and chromosomal abnormalities in vitro.
Specifikationer
Specifikationer
| Kemiskt namn eller material | NSC 617145 |
| CAS | 203115-63-3 |
| Synonym | 1,1'-(2,2-Dimethyl-1,3-propanediyl)bis[3,4-dichloro-1H-pyrrole-2,5-dione |
| Kvantitet | 10 mg |
| Löslighet | Soluble to 20mM in DMSO |
| Molekylformel | C13H10Cl4N2O4 |
| Formel vikt | 400.04 |
| Fysisk form | Solid |
| Produkttyp | Bioactive Small Molecule |
| Renhet | >98% |
Research Use Only. Not For Human or Veterinary Use