Learn More
Tocris Bioscience™ PFI 4
Potent and selective BRPF1 bromodomain inhibitor
Brand: Tocris Bioscience™ 5576/10
This item is not returnable.
View return policy
Description
PFI 4 is a potent and selective BRPF1B bromodomain inhibitor (IC50 = 172 nM). Exhibits >100-fold selectivity for BRPF1 over a panel of other bromodomains including BRPF2 (BRD1), BRPF3 and BRD4. Disrupts BRPF1 binding to histone H3.3. Cell permeable. Inhibits osteoclastogenesis.
Specifications
PFI 4 | |
N-[2,3-Dihydro-1,3-dimethyl-2-oxo-6-(1-pyrrolidinyl)-1H-benzimidazol-5-yl]-2-methoxybenzamide | |
Soluble to 50mM in DMSO and to 10mM in ethanol | |
380.44 | |
Bioactive Small Molecule |
900305-37-5 | |
10 mg | |
C21H24N4O3 | |
Solid | |
>99% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tocris Bioscience™ PFI 4 > 10mg
Research Use Only. Not For Human or Veterinary Use
Spot an opportunity for improvement?Share a Content Correction