Läs mer
(R)-De(trifluoromethyl) Fluoxetine Hydrochloride, TRC
Produktkod. 30349683
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30349683 100 mg
1 options
Produktkod. 30349683 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD298360100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-De(trifluoromethyl) Fluoxetine Hydrochloride
CAS 1212215-97-8
Molekylformel C16 H19 N O . Cl H
Kvantitet 100 mg
Synonym (γR)-N-Methyl-γ-phenoxybenzenepropanamine Hydrochloride (1:1), (R)-N-Methyl-3-phenoxy-3-phenylpropan-1-amine Hydrochloride, Benzenepropanamine, N-methyl-γ-phenoxy-, hydrochloride (1:1), (γR)-, (3R)-N-Methyl-3-phenoxy-3-phenylpropan-1-amine Hydrochloride
InChI formel InChI=1S/C16H19NO.ClH/c1-17-13-12-16(14-8-4-2-5-9-14)18-15-10-6-3-7-11-15;/h2-11,16-17H,12-13H2,1H3;1H/t16-;/m1./s1
LEDER Cl.CNCC[C@@H](Oc1ccccc1)c2ccccc2
IUPAC-namn (3R)-N-methyl-3-phenoxy-3-phenylpropan-1-amine;hydrochloride
Molekylvikt (g/mol) 277.79
Formel vikt 277.1233