Läs mer
R-(-)-Deprenyl N-Oxide Hydrochloride Salt, TRC
Produktkod. 30004753
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004753 10 mg
30006492 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30004753 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28864510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 830318-01-9
Molekylformel C13 H18 N O . Cl
Molekylvikt (g/mol) 239.74
Synonym (αR)-N,α-Dimethyl-N-2-propynylbenzene N-Oxide Hydrochloride Salt, (αR)-N,α-Dimethyl-N-2-propyn-1-yl-benzeneethanamine N-Oxide Hydrochloride Salt, Selegiline N-oxide hydrochloride
IUPAC-namn hydroxy-methyl-[(1R)-1-methyl-2-phenyl-ethyl]-prop-2-ynyl-ammonium;chloride
LEDER [Cl-].C[C@H](Cc1ccccc1)[N+](C)(O)CC#C
Specifikationer
Kemiskt namn eller material R-(-)-Deprenyl N-Oxide hydrochloride
Kvantitet 10 mg
InChI formel InChI=1S/C13H18NO.ClH/c1-4-10-14(3,15)12(2)11-13-8-6-5-7-9-13;/h1,5-9,12,15H,10-11H2,2-3H3;1H/q+1;/p-1/t12-,14?;/m1./s1
Formel vikt 239.1077