Läs mer
R-Dihydroactinidiolide, TRC
Produktkod. 30428535
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30428535 250 mg
1 options
Produktkod. 30428535 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR268145250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (-)-Dihydroactinidiolide
CAS 17092-92-1
Molekylformel C11 H16 O2
Kvantitet 250 mg
Synonym (-)-Dihydroactinidiolide, 2(4H)-Benzofuranone','5,6,7,7a-tetrahydro-4,4,7a-trimethyl-','(7aR)- (9CI','ACI), (7aR)-5,6,7,7a-Tetrahydro-4,4,7a-trimethyl-2(4H)-benzofuranone (ACI), 2(4H)-Benzofuranone','5,6,7,7a-tetrahydro-4,4,7a-trimethyl- (8CI), 2(4H)-Benzofur
InChI formel InChI=1S/C11H16O2/c1-10(2)5-4-6-11(3)8(10)7-9(12)13-11/h7H,4-6H2,1-3H3/t11-/m1/s1
LEDER CC1(C)CCC[C@@]2(C)OC(=O)C=C12
IUPAC-namn (7aR)-4,4,7a-trimethyl-6,7-dihydro-5H-1-benzofuran-2-one
Molekylvikt (g/mol) 180.244
Formel vikt 180.115