Läs mer
(R)-(+)-Dropropizine, TRC
Produktkod. 30135737
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135737 1 g
30004986 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135737 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD6815051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 99291-24-4
Molekylformel C13 H20 N2 O2
Molekylvikt (g/mol) 236.31
Synonym Levodropropizine impurity A, (2R)-3-(4-phenylpiperazin-1-yl)propane-1,2-diol, Dextrodropropizine
IUPAC-namn (2R)-3-(4-phenylpiperazin-1-yl)propane-1,2-diol
LEDER OC[C@H](O)CN1CCN(CC1)c2ccccc2
Specifikationer
Kemiskt namn eller material (2R)-3-(4-phenylpiperazin-1-yl)propane-1,2-diol
Kvantitet 1 g
InChI formel InChI=1S/C13H20N2O2/c16-11-13(17)10-14-6-8-15(9-7-14)12-4-2-1-3-5-12/h1-5,13,16-17H,6-11H2/t13-/m1/s1
Formel vikt 236.1525