Läs mer
(R)-Haloxyfop Methyl Ester, TRC
Produktkod. 30133049
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133049 250 mg
30137892 1 g
17961235 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133049 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH104020250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 72619-32-0
Molekylformel C16 H13 Cl F3 N O4
Molekylvikt (g/mol) 375.73
Synonym Propanoic acid, 2-[4-[[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy]phenoxy]-, methyl ester, (R)-, (R)-Haloxyfop methyl ester, (R)-Haloxyfop-methyl, DE 535, Edge, Eloge, Gallant super, Haloxyfop-P-methyl, Haloxyfop-R-methyl, Zellek Super
IUPAC-namn methyl (2R)-2-[4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxyphenoxy]propanoate
LEDER COC(=O)[C@@H](C)Oc1ccc(Oc2ncc(cc2Cl)C(F)(F)F)cc1
Specifikationer
Kemiskt namn eller material Haloxyfop-R-methyl
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C16H13ClF3NO4/c1-9(15(22)23-2)24-11-3-5-12(6-4-11)25-14-13(17)7-10(8-21-14)16(18,19)20/h3-9H,1-2H3/t9-/m1/s1
Formel vikt 375.0485
Procent renhet >95%