Läs mer
R&D Systems™ Human Erythropoietin R (NP_000112) VersaClone cDNA
Produktkod. 18713279
Change view
Kvantitet:
10 μg
Produktkod. Kvantitet
18713279 10 μg
1 options
Produktkod. 18713279 Leverantör R&D Systems™ Leverantörsnummer RDC1158
  • ONLINE EXCLUSIVE PRICE
    3500.00 SEK
    3309.60 SEK / 10µg
    Save 190.40 SEK
    5% Av
 Begränsad artikel
Beräknad leverans: 19-10-2026
Erythropoietin Receptor
The Human Erythropoietin R (NP_000112) VersaClone cDNA clone contains the complete ORF for Erythropoietin R, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Erythropoietin R insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Beställningsinfo
Ships at ambient temperature
TRUSTED_SUSTAINABILITY
Specifikationer
Innehåll och lagring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Kvantitet 10 μg
For Reasearch Use Only; Not for Use in Humans