Läs mer
R&D Systems™ Human LRRC33 (NP_940967) VersaClone cDNA
Produktkod. 18748849
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18748849 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Produktkod. 18748849 Leverantör R&D Systems™ Leverantörsnummer RDC1138
  • ONLINE EXCLUSIVE PRICE
    8080.00 SEK
    7641.90 SEK / 10µg
    Save 438.10 SEK
    5% Av
 Begränsad artikel
Beräknad leverans: 07-10-2026
Lägg i varukorgen
Lägg i varukorgen
Leucine-rich Repeat Containing 33
The Human LRRC33 (NP_940967) VersaClone cDNA clone contains the complete ORF for LRRC33, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The LRRC33 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Beställningsinfo
Ships at ambient temperature
TRUSTED_SUSTAINABILITY
Specifikationer
Innehåll och lagring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Kvantitet 10 μg
For Reasearch Use Only; Not for Use in Humans