Läs mer
(R)-Limonene 1,2-epoxide (Mixture of Diastereomers), TRC
Produktkod. 30480860
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30480860 250 mg
1 options
Produktkod. 30480860 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL461750250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-Limonene 1,2-epoxide (Mixture of Diastereomers)
CAS 203719-54-4
Molekylformel C10H16O
Kvantitet 250 mg
Synonym (4R)-1-Methyl-4-(1-methylethenyl)-7-oxabicyclo[4.1.0]heptane, (R)-Limonene Epoxide
InChI formel InChI=1S/C10H16O/c1-7(2)8-4-5-10(3)9(6-8)11-10/h8-9H,1,4-6H2,2-3H3/t8-,9?,10?/m1/s1
LEDER CC(=C)[C@@H]1CCC2(C)OC2C1
IUPAC-namn (3R)-6-methyl-3-prop-1-en-2-yl-7-oxabicyclo[4.1.0]heptane
Molekylvikt (g/mol) 152.23
Formel vikt 152.1201