Läs mer
(R)-Lisinopril Sodium Salt, TRC
Produktkod. 30465338
Change view
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30465338 5 mg
30473072 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30465338 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL4689805MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-Lisinopril Sodium Salt
CAS 1356019-70-9
Molekylformel C21H31N3NaO5
Kvantitet 5 mg
Synonym (R)-1-[N2-(1-Carboxy-3-phenylpropyl)-L-lysyl]L-proline Sodium, N2-[(1R)-1-Carboxy-3-phenylpropyl)-L-lysyl]L-proline Sodium
LEDER O=C(N1[C@H](C(O)=O)CCC1)[C@H](CCCCN)N[C@@H](C(O)=O)CCC2=CC=CC=C2.[Na]
Molekylvikt (g/mol) 428.48