missing translation for 'onlineSavingsMsg'
Läs mer

(R)-Lisinopril Sodium Salt, TRC

Product Code. 30473072
Change view
Click to view available options
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30473072 1 mg
30465338 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30473072 Supplier Toronto Research Chemicals Supplier No. TRCL4689801MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material (R)-Lisinopril Sodium Salt
CAS 1356019-70-9
Molekylformel C21H31N3NaO5
Kvantitet 1 mg
Synonym (R)-1-[N2-(1-Carboxy-3-phenylpropyl)-L-lysyl]L-proline Sodium, N2-[(1R)-1-Carboxy-3-phenylpropyl)-L-lysyl]L-proline Sodium
LEDER O=C(N1[C@H](C(O)=O)CCC1)[C@H](CCCCN)N[C@@H](C(O)=O)CCC2=CC=CC=C2.[Na]
Molekylvikt (g/mol) 428.48
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.