Läs mer
(R)-Lofexidine, TRC
Produktkod. 30471696
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30471696 1 mg
1 options
Produktkod. 30471696 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL4693951MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-Lofexidine
CAS 81447-78-1
Molekylformel C11 H12 Cl2 N2 O
Kvantitet 1 mg
Synonym 1H-Imidazole, 2-[(1R)-1-(2,6-dichlorophenoxy)ethyl]-4,5-dihydro-, 1H-Imidazole, 2-[1-(2,6-dichlorophenoxy)ethyl]-4,5-dihydro-, (R)-, 2-[(1R)-1-(2,6-Dichlorophenoxy)ethyl]-4,5-dihydro-1H-imidazole, (-)-Lofexidine, (R)-(-)-Lofexidine, Levlofexidine
InChI formel InChI=1S/C11H12Cl2N2O/c1-7(11-14-5-6-15-11)16-10-8(12)3-2-4-9(10)13/h2-4,7H,5-6H2,1H3,(H,14,15)/t7-/m1/s1
LEDER C[C@@H](Oc1c(Cl)cccc1Cl)C2=NCCN2
IUPAC-namn 2-[(1R)-1-(2,6-dichlorophenoxy)ethyl]-4,5-dihydro-1H-imidazole
Molekylvikt (g/mol) 259.13
Formel vikt 258.0327