Läs mer
R-Mannostatin A, Hydrochloride (~90%), TRC
Produktkod. 30475527
Change view
Kvantitet:
0.5 mg
2.5 mg
5 mg
Förpackningsstorlek:
0.5mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30475527 2.5 mg 2.5mg
30465374 0.5 mg 0.5mg
30470207 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30475527 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM1665552.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 134677-22-8
Molekylformel C6H13NO3S . HCl
Molekylvikt (g/mol) 215.7
Synonym (1S,2S,3S,4R,5S)-4-Amino-5-(methylthio)cyclopentane-1,2,3-triol Hydrochloride
IUPAC-namn (1S,2S,3S,4R,5S)-4-amino-5-methylsulfanylcyclopentane-1,2,3-triol;hydrochloride
LEDER Cl.CS[C@H]1[C@H](N)[C@H](O)[C@H](O)[C@@H]1O
Specifikationer
Kemiskt namn eller material R-Mannostatin A, Hydrochloride
Kvantitet 2.5 mg
InChI formel InChI=1S/C6H13NO3S.ClH/c1-11-6-2(7)3(8)4(9)5(6)10;/h2-6,8-10H,7H2,1H3;1H/t2-,3+,4+,5+,6+;/m1./s1
Formel vikt 215.0383