Läs mer
(R)-(+)-Menthofuran (>85%), TRC
Produktkod. 30011520
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011520 100 mg
17942645 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30011520 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM218861100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17957-94-7
Molekylformel C10H14O
Molekylvikt (g/mol) 150.22
Synonym (6R)-4,5,6,7-Tetrahydro-3,6-dimethylbenzofuran, (+)-Menthofuran, (6R)-3,6-Dimethyl-4,5,6,7-tetrahydro-1-benzofuran, (R)-3,6-Dimethyl-4,5,6,7-tetrahydrobenzofuran, (R)-Menthofuran
IUPAC-namn (6R)-3,6-dimethyl-4,5,6,7-tetrahydro-1-benzofuran
LEDER C[C@@H]1CCc2c(C)coc2C1
Specifikationer
Kemiskt namn eller material (R)-(+)-Menthofuran
Kvantitet 100 mg
InChI formel InChI=1S/C10H14O/c1-7-3-4-9-8(2)6-11-10(9)5-7/h6-7H,3-5H2,1-2H3/t7-/m1/s1
Formel vikt 150.1045