Läs mer
(R)-Mephenytoin, TRC
Produktkod. 30464938
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30464938 10 mg
1 options
Produktkod. 30464938 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM22499010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-Mephenytoin
CAS 71140-51-7
Molekylformel C12H14N2O2
Kvantitet 10 mg
Synonym 2,4-Imidazolidinedione, 5-ethyl-3-methyl-5-phenyl-, (5R)-, 2,4-Imidazolidinedione, 5-ethyl-3-methyl-5-phenyl-, (R)-, (5R)-5-Ethyl-3-methyl-5-phenyl-2,4-imidazolidinedione, (-)-5-Ethyl-3-methyl-5-phenylhydantoin, (-)-Mephenytoin, (-)-Mesantoin, (5R)-5-Ethyl-3-methyl-5-phenylimidazolidine-2,4-dione, (R)-Mephenytoin, l-Mephenytoin
InChI formel InChI=1S/C12H14N2O2/c1-3-12(9-7-5-4-6-8-9)10(15)14(2)11(16)13-12/h4-8H,3H2,1-2H3,(H,13,16)/t12-/m1/s1
LEDER CC[C@@]1(NC(=O)N(C)C1=O)c2ccccc2
Molekylvikt (g/mol) 218.25
Formel vikt 218.1055