Läs mer
(R)-(-)-Mepivacaine Hydrochloride, TRC
Produktkod. 30479642
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30479642 250 mg
1 options
Produktkod. 30479642 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM225070250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-(-)-Mepivacaine Hydrochloride
CAS 34333-72-7
Molekylformel C15 H22 N2 O . Cl H
Kvantitet 250 mg
Synonym (R)-(-)-Mepivacaine Hydrochloride, (2R)-N-(2,6-Dimethylphenyl)-1-methyl-2-piperidinecarboxamide Hydrochloride, (-)-Mepivacaine Hydrochloride, (R)-Mepivacaine Hydrochloride, D-(-)-Mepivacaine Hydrochloride, D-Mepivacaine Hydrochloride, (R)-N-(2,6-Dimethylphenyl)-1-methyl-2-piperidinecarboxamide Hydrochloride
InChI formel InChI=1S/C15H22N2O.ClH/c1-11-7-6-8-12(2)14(11)16-15(18)13-9-4-5-10-17(13)3;/h6-8,13H,4-5,9-10H2,1-3H3,(H,16,18);1H/t13-;/m1./s1
LEDER Cl.CN1CCCC[C@@H]1C(=O)Nc2c(C)cccc2C
IUPAC-namn (2R)-N-(2,6-dimethylphenyl)-1-methylpiperidine-2-carboxamide;hydrochloride
Molekylvikt (g/mol) 282.809
Formel vikt 282.15