Läs mer
(R)-Methyl 5-Oxopyrrolidine-2-carboxylate, TRC
Produktkod. 30465897
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30465897 10 g
1 options
Produktkod. 30465897 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM13798510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-Methyl 5-Oxopyrrolidine-2-carboxylate
CAS 64700-65-8
Molekylformel C6H9NO3
Kvantitet 10 g
Synonym (R)-5-(Methoxycarbonyl)-2-pyrrolidinone, (R)-Methyl 2-pyrrolidone-5-carboxylateMethyl (R)-5-oxopyrrolidine-2-carboxylate, Methyl (R)-pyroglutamate
InChI formel InChI=1S/C6H9NO3/c1-10-6(9)4-2-3-5(8)7-4/h4H,2-3H2,1H3,(H,7,8)/t4-/m1/s1
LEDER COC(=O)[C@H]1CCC(=O)N1
IUPAC-namn methyl (2R)-5-oxopyrrolidine-2-carboxylate
Molekylvikt (g/mol) 143.14
Formel vikt 143.0582