Läs mer
R-(-)-N-Demethyl Deprenyl, TRC
Produktkod. 30134278
Change view
Kvantitet:
1 mg
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30134278 5 mg -
30010017 2.5 mg 2.5mg
30009716 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134278 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2309205MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56862-28-3
Molekylformel C12 H15 N . Cl H
Molekylvikt (g/mol) 209.72
Synonym N-[(1R)-1-Methyl-2-phenylethyl]prop-2-yn-1-amine Hydrochloride, Desmethylselegiline Hydrochloride, (R)-N-(1-Phenylpropan-2-yl)prop-2-yn-1-amine hydrochloride
IUPAC-namn (2R)-1-phenyl-N-prop-2-ynylpropan-2-amine;hydrochloride
LEDER Cl.C[C@H](Cc1ccccc1)NCC#C
Specifikationer
Kemiskt namn eller material R-(-)-N-Demethyl Deprenyl
Kvantitet 5 mg
InChI formel InChI=1S/C12H15N.ClH/c1-3-9-13-11(2)10-12-7-5-4-6-8-12;/h1,4-8,11,13H,9-10H2,2H3;1H/t11-;/m1./s1
Formel vikt 209.0971