Läs mer
(R)-N-Desacryloyl N-3-Chloropropionyl Ibrutinib, TRC
Produktkod. 30365479
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30365479 50 mg
1 options
Produktkod. 30365479 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28811050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-N-Desacryloyl N-3-Chloropropionyl Ibrutinib
CAS 1288338-96-4
Molekylformel C25 H25 Cl N6 O2
Kvantitet 50 mg
Synonym (R)-N-Desacryloyl N-3-Propionyl Ibrutinib, 1-Propanone, 1-[(3R)-3-[4-amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl]-1-piperidinyl]-3-chloro- (ACI), 1-[(3R)-3-[4-Amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl]-1-piperidinyl]-3-chloro-1-propanone (ACI), (R)-1-[3-[4-Amino-3-(
InChI formel InChI=1S/C25H25ClN6O2/c26-13-12-21(33)31-14-4-5-18(15-31)32-25-22(24(27)28-16-29-25)23(30-32)17-8-10-20(11-9-17)34-19-6-2-1-3-7-19/h1-3,6-11,16,18H,4-5,12-15H2,(H2,27,28,29)/t18-/m1/s1
LEDER Nc1ncnc2c1c(nn2[C@@H]3CCCN(C3)C(=O)CCCl)c4ccc(Oc5ccccc5)cc4
IUPAC-namn 1-[(3R)-3-[4-amino-3-(4-phenoxyphenyl)pyrazolo[3,4-d]pyrimidin-1-yl]piperidin-1-yl]-3-chloropropan-1-one
Molekylvikt (g/mol) 476.958
Formel vikt 476.173