Läs mer
(R)-N-Formyl Octabase, TRC
Produktkod. 30010289
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30010289 50 mg
1 options
Produktkod. 30010289 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF70088850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-N-Formyl Octabase
CAS 51773-23-0
Molekylformel C18H23NO2
Kvantitet 50 mg
Synonym (R)-3,4,5,6,7,8-Hexahydro-1-[(4-methoxyphenyl)methyl]-2(1H)-isoquinolinecarboxaldehyde;
LEDER COc1ccc(C[C@H]2N(CCC3=C2CCCC3)C=O)cc1
Molekylvikt (g/mol) 285.38