Läs mer
(R)-(+)-Nor Verapamil Hydrochloride, TRC
Produktkod. 30424605
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30424605 1 mg
30421950 5 mg
30410782 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30424605 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN8774901MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 147158-71-2
Molekylformel C26 H36 N2 O4 . Cl H
Molekylvikt (g/mol) 477.04
Synonym Benzeneacetonitrile, α-[3-[[2-(3,4-dimethoxyphenyl)ethyl]amino]propyl]-3,4-dimethoxy-α-(1-methylethyl)-, monohydrochloride, (αR)- (9CI), Benzeneacetonitrile, α-[3-[[2-(3,4-dimethoxyphenyl)ethyl]amino]propyl]-3,4-dimethoxy-α-(1-methylethyl)-, monohydrochloride, (R)-, (R)-Norverapamil hydrochloride, LU 48855, (R)-(+)-Nor Verapamil Hydrochloride
IUPAC-namn (2R)-2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethylamino]-2-propan-2-ylpentanenitrile;hydrochloride
LEDER Cl.COc1ccc(CCNCCC[C@@](C#N)(C(C)C)c2ccc(OC)c(OC)c2)cc1OC
Specifikationer
Kemiskt namn eller material (R)-(+)-Norverapamil Hydrochloride
Kvantitet 1 mg
InChI formel InChI=1S/C26H36N2O4.ClH/c1-19(2)26(18-27,21-9-11-23(30-4)25(17-21)32-6)13-7-14-28-15-12-20-8-10-22(29-3)24(16-20)31-5;/h8-11,16-17,19,28H,7,12-15H2,1-6H3;1H/t26-;/m1./s1
Formel vikt 476.2442