Läs mer
(R)-(-)-Norlaudanosine Hydrochloride, TRC
Produktkod. 30428515
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30428515 100 mg
1 options
Produktkod. 30428515 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN661415100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-(-)-Norlaudanosine Hydrochloride
CAS 54417-53-7
Molekylformel C20H26ClNO4
Kvantitet 100 mg
Synonym (R)-1-(3,4-Dimethoxybenzyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline Hydrochloride, D)-(-)-Norlaudanosine Hydrochloride, (R)-Tetrahydropapaverine Hydrochloride, D-(-)-Tetrahydropapaverine Hydrochloride
InChI formel InChI=1S/C20H25NO4.ClH/c1-22-17-6-5-13(10-18(17)23-2)9-16-15-12-20(25-4)19(24-3)11-14(15)7-8-21-16;/h5-6,10-12,16,21H,7-9H2,1-4H3;1H/t16-;/m1./s1
LEDER Cl.COc1ccc(C[C@H]2NCCc3cc(OC)c(OC)cc23)cc1OC
IUPAC-namn (1R)-1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline;hydrochloride
Molekylvikt (g/mol) 379.88
Formel vikt 379.155