Läs mer
(R)-O-Desmethyl Naproxen, TRC
Produktkod. 30353224
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30353224 10 mg
1 options
Produktkod. 30353224 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29212410MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-O-Desmethyl Naproxen
CAS 123050-98-6
Molekylformel C13 H12 O3
Kvantitet 10 mg
Synonym 2-Naphthaleneacetic acid, 6-hydroxy-α-methyl-, (αR)- (9CI, ACI), (αR)-6-Hydroxy-α-methyl-2-naphthaleneacetic acid (ACI), 2-Naphthaleneacetic acid, 6-hydroxy-α-methyl-, (R)- (ZCI), (2R)-2-(6-Hydroxynaphthalen-2-yl)propanoic acid, (R)-2-(6-Hydroxynaphthalen-2-yl)propanoic acid, (R)-Naproxen-O-desmethyl, (R)-O-Demethylnaproxen
InChI formel InChI=1S/C13H12O3/c1-8(13(15)16)9-2-3-11-7-12(14)5-4-10(11)6-9/h2-8,14H,1H3,(H,15,16)/t8-/m1/s1
LEDER C[C@@H](C(=O)O)c1ccc2cc(O)ccc2c1
IUPAC-namn (2R)-2-(6-hydroxynaphthalen-2-yl)propanoic acid
Molekylvikt (g/mol) 216.23
Formel vikt 216.0786