Läs mer
(R)-Penbutolol Hydrochloride, TRC
Produktkod. 30135615
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135615 50 mg
30008971 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135615 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP22049050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57130-27-5
Molekylformel C18H29NO2.ClH
Molekylvikt (g/mol) 327.89
Synonym (R)-1-(2-cyclopentylphenoxy)-3-[(1,1-dimethylethyl)amino]-2-propanol Hydrochloride, (+)-(R)-Penbutolol Hydrochloride, (+)-Isopenbutolol Hydrochloride, (+)-Penbutolol Hydrochloride, Isopenbutolol Hydrochloride, d-Penbutolol Hydrochloride;
IUPAC-namn (2R)-1-(tert-butylamino)-3-(2-cyclopentylphenoxy)propan-2-ol;hydrochloride
LEDER Cl.CC(C)(C)NC[C@@H](O)COc1ccccc1C2CCCC2
Specifikationer
Kemiskt namn eller material (R)-Penbutolol Hydrochloride
Kvantitet 50 mg
InChI formel InChI=1S/C18H29NO2.ClH/c1-18(2,3)19-12-15(20)13-21-17-11-7-6-10-16(17)14-8-4-5-9-14;/h6-7,10-11,14-15,19-20H,4-5,8-9,12-13H2,1-3H3;1H/t15-;/m1./s1
Formel vikt 327.1965