Läs mer
R-Perindopril Diketopiperazine, TRC
Produktkod. 30420559
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30420559 25 mg
30424335 10 mg
30411371 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30420559 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP29764525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 925913-14-0
Molekylformel C19H30N2O4
Molekylvikt (g/mol) 350.45
Synonym Ethyl (alphaR,3S,5aS,9aS,10aS)-decahydro-3-methyl-1,4-dioxo-alpha-propylpyrazino[1,2-a]indole-2(1H)-acetate, (alphaR,3S,5aS,9aS,10aS)-Pyrazino[1,2-a]indole-2(1H)-acetic acid, decahydro-3-methyl-1,4-dioxo-alpha-propyl-ethyl Ester
IUPAC-namn ethyl (2R)-2-[(3S,5aS,9aS,10aS)-3-methyl-1,4-dioxo-5a,6,7,8,9,9a,10,10a-octahydro-3H-pyrazino[1,2-a]indol-2-yl]pentanoate
LEDER CCC[C@@H](N1[C@@H](C)C(=O)N2[C@H]3CCCC[C@H]3C[C@H]2C1=O)C(=O)OCC
Specifikationer
Kemiskt namn eller material R-Perindopril Diketopiperazine
Kvantitet 25 mg
InChI formel InChI=1S/C19H30N2O4/c1-4-8-15(19(24)25-5-2)20-12(3)17(22)21-14-10-7-6-9-13(14)11-16(21)18(20)23/h12-16H,4-11H2,1-3H3/t12-,13-,14-,15+,16-/m0/s1
Formel vikt 350.2206