Läs mer
(R)-Pinocembrin, TRC
Produktkod. 30413171
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30413171 500 mg
30428348 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30413171 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP468505500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 206660-42-6
Molekylformel C15H12O4
Molekylvikt (g/mol) 256.25
Synonym (2R)-2,3-Dihydro-5,7-dihydroxy-2-phenyl-4H-1-benzopyran-4-one, (R)-2,3-dihydro-5,7-dihydroxy-2-phenyl-4H-1-benzopyran-4-one, (-)-Pinocembrin, (-)-Pinocoembrin
IUPAC-namn (2R)-5,7-dihydroxy-2-phenyl-2,3-dihydrochromen-4-one
LEDER C1=CC=C(C=C1)[C@H]2CC(=O)C3=C(C=C(C=C3O2)O)O
Specifikationer
Kemiskt namn eller material (R)-Pinocembrin
Kvantitet 500 mg
InChI formel InChI=1S/C15H12O4/c16-10-6-11(17)15-12(18)8-13(19-14(15)7-10)9-4-2-1-3-5-9/h1-7,13,16-17H,8H2/t13-/m1/s1
Formel vikt 256.0736