Läs mer
(R)-Praziquantel, TRC
Produktkod. 30421277
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30421277 100 mg
1 options
Produktkod. 30421277 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP702100100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-Praziquantel
CAS 57452-98-9
Molekylformel C19H24N2O2
Kvantitet 100 mg
Synonym (11bR)-2-(Cyclohexylcarbonyl)-1,2,3,6,7,11b-hexahydro-4H-Pyrazino[2,1-a]isoquinolin-4-one, (R)-2-(Cyclohexylcarbonyl)-1,2,3,6,7,11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one, (-)-Praziquantel, (R)-(-)-Praziquantel, l-Praziquantel
InChI formel InChI=1S/C19H24N2O2/c22-18-13-20(19(23)15-7-2-1-3-8-15)12-17-16-9-5-4-6-14(16)10-11-21(17)18/h4-6,9,15,17H,1-3,7-8,10-13H2/t17-/m0/s1
LEDER O=C(C1=CC=C(OCCN2CCCCC2)C=C1)C3=C(C4=CC=C(O[C@@H]5O[C@H](C(O[Li])=O)[C@@H](O)[C@H](O)[C@H]5O)C=C4)SC6=C3C=CC(O)=C6
IUPAC-namn (11bR)-2-(cyclohexanecarbonyl)-3,6,7,11b-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4-one
Molekylvikt (g/mol) 312.41
Formel vikt 312.1838