Läs mer
(R)-Pregabalin, Mikromol™
Produktkod. 17942766
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17942766 100 mg
17960766 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17942766 Leverantör Mikromol Leverantörsnummer MM1376.02
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(R)-Pregabalin
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-Pregabalin
CAS 148553-51-9
Analyt- eller komponentnamn (R)-Pregabalin
Frakt skick Room Temperature
Molekylformel C8 H17 N O2
Certifikat och efterlevnad ISO 17025
Synonym Pregabalin Imp. B (EP), Hexanoic acid, 3-(aminomethyl)-5-methyl-, (3R)-, Hexanoic acid, 3-(aminomethyl)-5-methyl-, (R)-, (R)-3-(Aminomethyl)-5-methylhexanoic acid, (R)-Pregabalin, PD 144550, ent-Pregabalin, Pregabalin Related Compound A, Pregabalin Impurity B
InChI formel InChI=1S/C8H17NO2/c1-6(2)3-7(5-9)4-8(10)11/h6-7H,3-5,9H2,1-2H3,(H,10,11)/t7-/m1/s1
LEDER CC(C)C[C@@H](CN)CC(=O)O
IUPAC-namn (3R)-3-(aminomethyl)-5-methylhexanoic acid
Molekylvikt (g/mol) 159.23
Kvantitet 100 mg
Formel vikt 159.1259 g/mol