Learn More
R&D Systems™ Quantikine Immunoassay Control Set 826 for Human ErbB2/Her2
Product Code. 30001945
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
Product Code. Quantity Unit Size
30001945 1 Each Each
1 options
Product Code. 30001945 Supplier R&D Systems™ Supplier No. QC86
  • ONLINE EXCLUSIVE PRICE
    2282.89 SEK
    2013.90 SEK / Paket med 1
    Save 268.99 SEK
    11% Av
 Restricted Item
Beräknad leverans: 09-12-2026
For use with catalog number DHER20
These lyophilized controls are designed for use as quantitative controls for determining cytokine concentrations in biological fluids.
  • Analyte: Human ErbB2/Her2
  • Form: Lyophilized
  • Composition: Diluted porcine serum with preservatives
  • Concentration Levels: Low, medium, and high
  • Storage: Unreconstituted controls should be stored at 2°C to 8°C and are stable for at least 6 months from receipt.
  • Usage: For research use only
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes ErbB2/Her2
Sample Type Biological Fluids
Type Immunoassay Control
Gene ID (Entrez) 2064
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Human
Form Lyophilized