Läs mer
(R)-Quizalofop-tefuryl, TRC
Produktkod. 30134997
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134997 25 mg
30138218 100 mg
30004510 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134997 Leverantör Toronto Research Chemicals Leverantörsnummer TRCQ82610525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 200509-41-7
Molekylformel C22 H21 Cl N2 O5
Molekylvikt (g/mol) 428.87
Synonym C 4874, Pantera, Pantera 45EC, Quizalofop P tefuryl, Quizalofop-P-tefuryl, UBI-C 4874, Propanoic acid, 2-[4-[(6-chloro-2-quinoxalinyl)oxy]phenoxy]-, (tetrahydro-2-furanyl)methyl ester, (2R)- (9CI, ACI)
IUPAC-namn oxolan-2-ylmethyl (2R)-2-[4-(6-chloroquinoxalin-2-yl)oxyphenoxy]propanoate
LEDER C[C@@H](Oc1ccc(Oc2cnc3cc(Cl)ccc3n2)cc1)C(=O)OCC4CCCO4
Specifikationer
Kemiskt namn eller material Pantera
Kvantitet 25 mg
InChI formel InChI=1S/C22H21ClN2O5/c1-14(22(26)28-13-18-3-2-10-27-18)29-16-5-7-17(8-6-16)30-21-12-24-20-11-15(23)4-9-19(20)25-21/h4-9,11-12,14,18H,2-3,10,13H2,1H3/t14-,18?/m1/s1
Formel vikt 428.1139