Läs mer
(R,R)-Dihydro Bupropion Hydrochloride, TRC
Produktkod. 30138478
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138478 50 mg
30003990 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138478 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44827550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 357637-18-4
Molekylformel C13 H20 Cl N O . HCl
Molekylvikt (g/mol) 278.22
Synonym (alphaR)-3-Chloro-alpha-[(1R)-1-[(1,1-dimethylethyl)amino]ethyl]-benzenemethanol Hydrochloride
IUPAC-namn (1R,2R)-2-(tert-butylamino)-1-(3-chlorophenyl)propan-1-ol;hydrochloride
LEDER Cl.C[C@@H](NC(C)(C)C)[C@H](O)c1cccc(Cl)c1
Specifikationer
Kemiskt namn eller material (R,R)-Dihydro Bupropion Hydrochloride
Kvantitet 50 mg
InChI formel InChI=1S/C13H20ClNO.ClH/c1-9(15-13(2,3)4)12(16)10-6-5-7-11(14)8-10;/h5-9,12,15-16H,1-4H3;1H/t9-,12+;/m1./s1
Formel vikt 277.1