Läs mer
(R,R)-Labetalol Hydrochloride, TRC
Produktkod. 30137021
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137021 10 mg
30007088 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137021 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL09650010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 75659-08-4
Molekylformel C19 H24 N2 O3 . Cl H
Molekylvikt (g/mol) 364.87
Synonym Benzamide, 2-hydroxy-5-[(1R)-1-hydroxy-2-[[(1R)-1-methyl-3-phenylpropyl]amino]ethyl]-, hydrochloride (1:1), Benzamide, 2-hydroxy-5-[(1R)-1-hydroxy-2-[[(1R)-1-methyl-3-phenylpropyl]amino]ethyl]-, monohydrochloride (9CI), Benzamide, 2-hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]-, monohydrochloride, [R-(R*,R*)]-, (R,R)-Labetalol hydrochloride, Dilevalol hydrochloride, Sch 19927 HCl
IUPAC-namn 2-hydroxy-5-[(1R)-1-hydroxy-2-[[(2R)-4-phenylbutan-2-yl]amino]ethyl]benzamide;hydrochloride
LEDER Cl.C[C@H](CCc1ccccc1)NC[C@H](O)c2ccc(O)c(c2)C(=O)N
Specifikationer
Kemiskt namn eller material (R,R)-Labetalol hydrochloride
Kvantitet 10 mg
InChI formel InChI=1S/C19H24N2O3.ClH/c1-13(7-8-14-5-3-2-4-6-14)21-12-18(23)15-9-10-17(22)16(11-15)19(20)24;/h2-6,9-11,13,18,21-23H,7-8,12H2,1H3,(H2,20,24);1H/t13-,18+;/m1./s1
Formel vikt 364.1554