Läs mer
(R,R,R)-2-Azabicyclo[3.3.0]octane-3-carboxylic Acid Benzyl Ester Hydrochloride Salt, TRC
Produktkod. 30334428
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30334428 100 mg
1 options
Produktkod. 30334428 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA795280100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R,R,R)-2-Azabicyclo[3.3.0]octane-3-carboxylic Acid Benzyl Ester Hydrochloride Salt
CAS 138877-09-5
Molekylformel C15H19NO2 . HCl
Kvantitet 100 mg
Synonym [2R-(2alpha,3abeta,6abeta)]-Octahydro-cyclopenta[b]pyrrole-2-carboxylic Acid Phenylmethyl Ester Hydrochloride
InChI formel InChI=1S/C15H19NO2.ClH/c17-15(18-10-11-5-2-1-3-6-11)14-9-12-7-4-8-13(12)16-14;/h1-3,5-6,12-14,16H,4,7-10H2;1H/t12-,13-,14-;/m1./s1
LEDER Cl.O=C(OCc1ccccc1)[C@H]2C[C@H]3CCC[C@H]3N2
IUPAC-namn benzyl (2R,3aR,6aR)-1,2,3,3a,4,5,6,6a-octahydrocyclopenta[b]pyrrole-2-carboxylate;hydrochloride
Molekylvikt (g/mol) 281.78
Formel vikt 281.1183