Läs mer
(R)-Rabeprazole Sodium Salt, TRC
Produktkod. 30427434
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30427434 100 mg
1 options
Produktkod. 30427434 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR070520100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-Rabeprazole Sodium Salt
CAS 171440-18-9
Molekylformel C18H20N3NaO3S
Kvantitet 100 mg
Synonym 2-[(R)-[[4-(3-Methoxypropoxy)-3-methyl-2-pyridinyl]methyl]sulfinyl]-1H-benzimidazole Sodium Salt, R-(+)-Rabeprazole Sodium Salt
InChI formel InChI=1S/C18H20N3O3S.Na/c1-13-16(19-9-8-17(13)24-11-5-10-23-2)12-25(22)18-20-14-6-3-4-7-15(14)21-18;/h3-4,6-9H,5,10-12H2,1-2H3;/q-1;+1/t25-;/m1./s1
LEDER O=[S@](CC1=NC=CC(OCCCOC)=C1C)C2=NC3=CC=CC=C3N2[Na]
IUPAC-namn sodium;2-[(R)-[4-(3-methoxypropoxy)-3-methylpyridin-2-yl]methylsulfinyl]benzimidazol-1-ide
Molekylvikt (g/mol) 381.42
Formel vikt 381.1123