Läs mer
(R)-Rivastigmine, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | (R)-Rivastigmine tartrate |
| CAS | 415973-05-6 |
| Molekylformel | C14 H22 N2 O2 . C4 H6 O6 |
| Kvantitet | 5 mg |
| Synonym | (R)-3-[1-(Dimethylamino)ethyl]phenyl ethylmethylcarbamate, hydrogen tartrate, Rivastigmine Tartrate R-Isomer, Rivastigmine Imp. D (EP), 3-[(1R)-1-(Dimethylamino)ethyl]phenyl N-ethyl-N-methylcarbamate hydrogen tartrate, (R)-Rivastigmine hydrogen tartrate, (R)-Rivastigmine tartrate |
| InChI formel | InChI=1S/C14H22N2O2.C4H6O6/c1-6-16(5)14(17)18-13-9-7-8-12(10-13)11(2)15(3)4;5-1(3(7)8)2(6)4(9)10/h7-11H,6H2,1-5H3;1-2,5-6H,(H,7,8)(H,9,10)/t11-;1-,2-/m11/s1 |
| LEDER | CCN(C)C(=O)Oc1cccc(c1)[C@@H](C)N(C)C.O[C@H]([C@@H](O)C(=O)O)C(=O)O |
| IUPAC-namn | (2R,3R)-2,3-dihydroxybutanedioic acid;[3-[(1R)-1-(dimethylamino)ethyl]phenyl] N-ethyl-N-methylcarbamate |
| Molekylvikt (g/mol) | 400.42 |
| Formel vikt | 400.1846 |