Läs mer
(R)-RuCl[(p-cymene)(BINAP)]Cl, TRC
Produktkod. 30409043
Change view
Kvantitet:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30409043 50 mg
30421738 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30409043 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR70157650MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-RuCl[(p-cymene)(BINAP)]Cl
CAS 145926-28-9
Molekylformel C45 H35 Cl P2 Ru . C10 H14 . Cl
Kvantitet 50 mg
Synonym Chloro[(R)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) Chloride
InChI formel InChI=1S/C44H32P2.C10H14.CH3.2ClH.Ru/c1-5-19-35(20-6-1)45(36-21-7-2-8-22-36)41-31-29-33-17-13-15-27-39(33)43(41)44-40-28-16-14-18-34(40)30-32-42(44)46(37-23-9-3-10-24-37)38-25-11-4-12-26-38;1-8(2)10-6-4-9(3)5-7-10;;;;/h1-32H;4-8H,1-3H3;1H3;2*1H;/q;;;;;+2/p-2
LEDER [Cl-].CC(C)c1ccc(C)cc1.C[Ru+]2(Cl)[P](c3ccccc3)(c4ccccc4)c5ccc6ccccc6c5c7c(ccc8ccccc78)[P]2(c9ccccc9)c%10ccccc%10
Molekylvikt (g/mol) 943.901
Formel vikt 943.173