Läs mer
(R,S)-Alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic Acid, TRC
Produktkod. 30137431
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137431 100 mg
30138639 50 mg
30004906 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137431 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA611500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 77521-29-0
Molekylformel C7 H10 N2 O4
Molekylvikt (g/mol) 186.17
Synonym alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, α-Amino-2,3-dihydro-5-methyl-3-oxo-4-isoxazolepropanoic acid, (R,S)-α-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, (RS)-α-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, (±)-α-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, 2-Amino-3-(3-hydroxy-5-methyl-1,2-oxazol-4-yl)propanoic acid, 2-Amino-3-(5-methyl-3-oxo-2,3-dihydro-1,2-oxazol-4-yl)propanoic acid, 2-Amino-3-(5-methyl-3-oxo-2,3-dihydroisoxazol-4-yl)propanoic acid, AMPA, AMPA (pharmaceutical), D,L-α-Amino-3-hydroxy-5-methylisoxazole-4-propionic acid, dl-α-Amino-3-hydroxy-5-methylisoxazole-4-propionic acid, α-Amino-2,3-dihydro-5-methyl-3-oxoisoxazole-4-propionic acid, α-Amino-3-hydroxy-5-methyl-4-isoxazolepropionate, α-Amino-3-hydroxy-5-methylisoxazole-4-propionic acid, γ-Amino-3-hydroxy-5-methylisoxazole-4-propionic acid
IUPAC-namn 2-amino-3-(3-hydroxy-5-methyl-1,2-oxazol-4-yl)propanoic acid
LEDER Cc1onc(O)c1CC(N)C(=O)O
Specifikationer
Kemiskt namn eller material (r,s)-a-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic Acid
Kvantitet 100 mg
InChI formel InChI=1S/C7H10N2O4/c1-3-4(6(10)9-13-3)2-5(8)7(11)12/h5H,2,8H2,1H3,(H,9,10)(H,11,12)
Formel vikt 186.0641