Läs mer
(R*,S*)-(±)-Fenoterol Hydrobromide, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 5-[(1RS)-2-[(1SR)-2-(4-Hydroxyphenyl)-1-methylethyl]amino-1-hydroxyethyl]benzene-1,3-diol Hydrobromide |
| CAS | 38964-10-2 |
| Molekylformel | C17 H21 N O4 . Br H |
| Kvantitet | 5 mg |
| Synonym | (R*,S*)-(±)-Fenoterol Hydrobromide, rel-5-[(1S)-1-Hydroxy-2-[[(1R)-2-(4-hydroxyphenyl)-1-methylethyl]amino]ethyl]-1,3-benzenediol Hydrobromide (1:1), 1,3-Benzenediol, 5-[1-hydroxy-2-[[2-(4-hydroxyphenyl)-1-methylethyl]amino]ethyl]-, hydrobromide, (R*,S*)- (9CI), 1,3-Benzenediol, 5-[1-hydroxy-2-[[2-(4-hydroxyphenyl)-1-methylethyl]amino]ethyl]-, hydrobromide, (R*,S*)-(±)-, 5-[(1RS)-2-[(1SR)-2-(4-Hydroxyphenyl)-1-methylethyl]amino-1-hydroxyethyl]benzene-1,3-diol hydrobromide, rel-(R,S)-Fenoterol hydrobromide, (RS,SR)-Fenoterol hydrobromide |
| InChI formel | InChI=1S/C17H21NO4.BrH/c1-11(6-12-2-4-14(19)5-3-12)18-10-17(22)13-7-15(20)9-16(21)8-13;/h2-5,7-9,11,17-22H,6,10H2,1H3;1H/t11-,17-;/m0./s1 |
| LEDER | Br.C[C@@H](Cc1ccc(O)cc1)NC[C@H](O)c2cc(O)cc(O)c2 |
| IUPAC-namn | 5-[(1R)-1-hydroxy-2-[[(2S)-1-(4-hydroxyphenyl)propan-2-yl]amino]ethyl]benzene-1,3-diol;hydrobromide |
| Molekylvikt (g/mol) | 384.265 |
| Formel vikt | 383.073 |