Läs mer
(R,S)-N’-Nitrosonornicotine-13C6, TRC
Produktkod. 30476787
Change view
Kvantitet:
0.5 mg
1 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30476787 10 mg
30470044 0.5 mg
30467532 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30476787 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN53502510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2726380-44-3
Molekylformel C313C6H11N3O
Molekylvikt (g/mol) 183.16
Synonym 3-(1-Nitroso-2-pyrrolidinyl)pyridine-13C6, NNN-13C6
IUPAC-namn 3-(1-nitrosopyrrolidin-2-yl-2-13C)pyridine-2,3,4,5,6-13C5
LEDER O=NN(CCC1)[13CH]1[13C]2=[13CH][13CH]=[13CH]N=[13CH]2
Specifikationer
Kemiskt namn eller material (R,S)-N'-Nitrosonornicotine-13C6
Kvantitet 10 mg
InChI formel InChI=1S/C9H11N3O/c13-11-12-6-2-4-9(12)8-3-1-5-10-7-8/h1,3,5,7,9H,2,4,6H2/i1+1,3+1,5+1,7+1,8+1,9+1
Formel vikt 183.11