Läs mer
(R)-Selisistat, MedChemExpress
Produktkod. 16459117
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16459117 5 mg
16416538 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16459117 Leverantör Medchem Express Leverantörsnummer HY15452B
MedChemExpress (R)-Selisistat ((R)-EX-527) is a R-enantiomer of Selisistat. Selisistat (EX-527) is a potent and selective SIRT1 inhibitor with IC50 of 98 nM.
(R)-Selisistat ((R)-EX-527) is a R-enantiomer of Selisistat. Selisistat (EX-527) is a potent and selective SIRT1 inhibitor with IC50 of 98 nM.
Specifikationer
Kemiskt namn eller material (R)-Selisistat
Fysisk form Solid
CAS 848193-69-1
Färg Light Yellow
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C13H13ClN2O
För användning med (applikation) Cancer-programmed cell death
Synonym (R)-EX-527
Löslighetsinformation DMSO : 100 mg/mL (402.07 mM; Need ultrasonic)
LEDER O=C([C@H]1C(NC2=C3C=C(Cl)C=C2)=C3CCC1)N
Molekylvikt (g/mol) 248.71
Formel vikt 248.71
Procent renhet 96.84%