Läs mer
(R)-Simurosertib, MedChemExpress
Produktkod. 16493208
Encompass_Preferred
Change view
Kvantitet:
2 mg
5 mg
10 mg
50 mg
Förpackningsstorlek:
10mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16493208 10 mg 10mg
16489756 50 mg -
16450046 2 mg -
16405947 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16493208 Leverantör Medchem Express Leverantörsnummer HY100888A
MedChemExpress (R)-Simurosertib ((R)-TAK-931) is the (R)-enantiomer of Simurosertib. Simurosertib (TAK-931) is an orally active, selective and ATP-competitive cell division cycle 7 (CDC7) kinase inhibitor, with an IC50 of <0.3 nM .
(R)-Simurosertib ((R)-TAK-931) is the (R)-enantiomer of Simurosertib. Simurosertib (TAK-931) is an orally active, selective and ATP-competitive cell division cycle 7 (CDC7) kinase inhibitor, with an IC50 of <0.3 nM .
Specifikationer
Kemiskt namn eller material (R)-Simurosertib
Fysisk form Solid
CAS 1330782-69-8
Färg Light Yellow
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C17H19N5OS
För användning med (applikation) Neuroscience-Neuromodulation
Synonym (R)-TAK-931
Löslighetsinformation DMSO : 75 mg/mL (219.66 mM; Need ultrasonic)
LEDER O=C(C1=C(N2)C=C(C3=CNN=C3C)S1)N=C2[C@H]4CC5CCN4CC5
Molekylvikt (g/mol) 341.43
Formel vikt 341.43
Procent renhet 98.07%