Läs mer
R-Spondin 2 Antibody (879712R) [DyLight 680], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB32662Y
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
R-Spondin 2 Monoclonal antibody specifically detects R-Spondin 2 in Human samples. It is validated for ELISA
Specifikationer
| R-Spondin 2 | |
| Monoclonal | |
| DyLight 680 | |
| CRISTIN2, hRspo2, MGC35555, MGC43342, Roof plate-specific spondin-2, R-spondin 2 homolog (Xenopus laevis), R-spondin-2 | |
| Mouse myeloma cell line NS0-derived recombinant human R-Spondin 2, Met1-Gly205, Accession # Q6UXX9 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 879712R | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 340419 | |
| Store at 4°C in the dark. | |
| IgG1 |