Läs mer
R-Spondin 2 Antibody (879715R) [Allophycocyanin], Novus Biologicals™
Handla allt Bio Techne ProdukterBeskrivning
R-Spondin 2 Monoclonal antibody specifically detects R-Spondin 2 in Human samples. It is validated for ELISA
Specifikationer
Specifikationer
| Antigen | R-Spondin 2 |
| Användningsområden | ELISA |
| Klassificering | Monoclonal |
| Klona | 879715R |
| Konjugera | APC |
| Formulering | PBS |
| Gene Alias | CRISTIN2, hRspo2, MGC35555, MGC43342, Roof plate-specific spondin-2, R-spondin 2 homolog (Xenopus laevis), R-spondin-2 |
| Värd art | Rat |
| Immunogen | Mouse myeloma cell line NS0-derived recombinant human R-Spondin 2, Met1-Gly205, Accession # Q6UXX9 |
| Reningsmetod | Protein A or G purified from cell culture supernatant |
| Visa mer |