Läs mer
R-Stearoyl Carnitine Chloride, TRC
Produktkod. 30424720
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30424720 2.5 g 2.5g
30410289 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30424720 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6865152.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 32350-57-5
Molekylformel C25H50ClNO4
Molekylvikt (g/mol) 464.12
Synonym 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[(1-oxooctadecyl)oxy]-, chloride, (2R)- (9CI), 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[(1-oxooctadecyl)oxy]-, chloride, (R)- (ZCI), Ammonium, (3-carboxy-2-hydroxypropyl)trimethyl-, chloride, stearate, L- (8CI), Stearic acid, ester with (3-carboxy-2-hydroxypropyl)trimethylammonium chloride, L- (8CI)
IUPAC-namn (R)-3-carboxy-N,N,N-trimethyl-2-(stearoyloxy)propan-1-aminium chloride
LEDER O=C(CCCCCCCCCCCCCCCCC)O[C@H](CC(O)=O)C[N+](C)(C)C.[Cl-]
Specifikationer
Kemiskt namn eller material Stearoyl-L-carnitine Chloride
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 g
InChI formel InChI=1S/C25H49NO4.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-25(29)30-23(21-24(27)28)22-26(2,3)4;/h23H,5-22H2,1-4H3;1H/t23-;/m1./s1
Formel vikt 463.34
Procent renhet >95%