Läs mer
(R)-Styrene Glycol, TRC
Produktkod. 30408812
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30408812 25 g
1 options
Produktkod. 30408812 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS68781525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-Styrene Glycol
CAS 16355-00-3
Molekylformel C8H10O2
Kvantitet 25 g
Synonym (-)-1-Phenylethane-1,2-diol, (-)-1-Phenylethylene Glycol, (-)-Phenyl Glycol, (-)-Phenyl-1,2-ethanediol, (-)-Styrene Glycol, (1R)-1-Phenyl-1,2-ethanediol, (R)-(-)-1-Phenylethane-1,2-diol, (R)-1-Phenyl-1,2-ethanediol, (R)-2-Phenyl-1,2-ethanediol, (R)-(-)-1-Phenyl-1,2-ethanediol
InChI formel InChI=1S/C8H10O2/c9-6-8(10)7-4-2-1-3-5-7/h1-5,8-10H,6H2/t8-/m0/s1
LEDER OC[C@H](O)c1ccccc1
IUPAC-namn (1R)-1-phenylethane-1,2-diol
Molekylvikt (g/mol) 138.16
Formel vikt 138.0681